Interested in a bulk or custom version of this product? Request pricing.

IL1R1 Recombinant Human Protein, hIgG1-Fc Tag

Catalog number: 10126H02H25

 Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Interleukin 1 receptor, type I (IL1R1), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in cell biology research applications. It is suitable for use as protein standard or in other research applications.

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Thr 332) of human IL1R1 (NP_000868.1) fused to the human IgG1 Fc region at the C-terminus.

Activity:
Measured by its binding ability in a functional ELISA:
1. Immobilized human IL1B at 20 µg/ml(100 µl/well) can bind human IL1R1-Fc with a linear range of 6.4-160 ng/ml.
2. Immobilized human IL1B-His at 10 µg/ml(100 µl/well) can bind human IL1R1-Fc. The ED50 of human IL1R1-Fc is 0.198 µg/ml.
3. Immobilized human IL1B at 10 µg/ml(100 µl/well) can bind human IL1R1-Fc. The ED50 of human IL1R1-Fc is 0.48 µg/ml.

Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80.

Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2 mg/ml or lower. It is recommended that the protein be aliquoted and be used as soon as possible. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.

Expiration Date: Expires one year from date of receipt when stored as instructed.



This protein is manufactured by Sino Biological Inc.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Research Category: Immunology, Inflammation, Signal Transduction
Endotoxin Level: < 1 EU/µg
Expression: Mammalian
Form: Lyophilized
Gene Aliases: CD121A, D2S1473, IL-1R-alpha, IL1R, IL1RA, P80, IL1R1
Gene ID: 3554
Gene Symbol: IL1R1
Molecular Weight: 63 kDa
Product Line: Sino Biological®
Product Size: 5 x 5 µg
Protein Family: CD Proteins, Cytokines & Receptors
Protein Form: Recombinant, Truncated
Protein Name: Interleukin 1 receptor, type I
Protein Subtype: Cytokine Receptors, Interleukins
Protein Tag or Fusion: IgG1 Fc Tag
Purity: ≥ 85% by SDS-PAGE
Species: Human
Validated Application: Assay Standard
Shipping Condition: Room Temperature

Contents & storage

Store at -20 to -70 °C

Documents

Manuals & protocols

Recommended products