Interested in a bulk or custom version of this product? Request pricing.

L1CAM (CD171) Recombinant Human Protein, His Tag

Catalog number: 10140H08H5

 Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

L1 cell adhesion molecule (L1CAM/CD171), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Glu 1120) of human L1CAM (NP_000416.1) fused to a polyhistidine tag at the C-terminus.

Activity: This recombinant protein has not been tested.

Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80.

Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2 mg/ml or lower. It is recommended that the protein be aliquoted and be used as soon as possible. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.

Expiration Date: Expires one year from date of receipt when stored as instructed.



This protein is manufactured by Sino Biological Inc.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Protein Family: CD Proteins
Research Category: Biomarker-Related Pathway(s), Extracellular Matrix & Cell Adhesion, Neurobiology, Oncology
Endotoxin Level: < 1 EU/µg
Expression: Mammalian
Form: Lyophilized
Gene Aliases: L1CAM, CAML1, CD171, HSAS, HSAS1, MASA, MIC5, N-CAML1, S10, SPG1
Gene ID: 3897
Gene Symbol: L1CAM
Molecular Weight: 125 kDa
Product Line: Sino Biological®
Product Size: 5 µg
Protein Form: Cell Surface Protein, Recombinant, Truncated
Protein Name: L1 cell adhesion molecule
Protein Subtype: Glycoproteins
Protein Tag or Fusion: His Tag
Purity: ≥ 85% by SDS-PAGE
Species: Human
Validated Application: Assay Standard
Shipping Condition: Room Temperature

Contents & storage

Store at -20 to -70 °C

Documents

Manuals & protocols