MAX Efficiency® Stbl2™ Competent Cells

Catalog number: 10268019

Invitrogen™  Related applications: Transformation

Error loading your content!

  Catalog number
Select a plan
Unit size
List price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

MAX Efficiency® Stbl2™ Competent Cells are high-efficiency chemically competent cells specifically designed for cloning unstable inserts. In addition to recA1, a unique set of genetic markers allow for stable cloning of direct repeat and retroviral sequences and tandem array genes. MAX Efficiency® Stbl2™ Competent Cells offer:

• >1 x 109 transformants/µg plasmid DNA for cloning rare sequences
• Elimination of mcrBC-hsdRMS-mrr and mutation of mcrA—allows cloning of methylated genomic sequences
• A unique genotype that stabilizes direct repeat and retroviral sequences such as SIV and HIV
• Greatly increased plasmid yield and quality due to endA1 mutation
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Competent Cell Type: Chemically Competent
Species: E. coli
Format: Tube(s)
Transformation Efficiency: > 1x10^9
Bacterial or Yeast Strain: Stbl2™
Plasmid Type: May be used for plasmids > 20 kb
Product Size: 5 x 200 µL
Blue-White Screening: No
Cloning Unstable DNA: Suitable for cloning unstable DNA
Reduces Recombination: Yes
Cloning Methylated DNA: Yes
Improves Plasmid Quality: Yes
Propagating ccdB Vectors: Not for ccdB vector propagation
F' Episome (to make ssDNA): Lacks F' Episome
Preparing Unmethylated DNA: Not Suitable for preparing unmethylated DNA
T1 Phage - Resistant (tonA): No
High Throughput Compatibility: Not High Throughput-Compatible (Manual)
Shipping Condition: Dry Ice

Contents & storage

Contains:
• MAX Efficiency® Stbl2™ Competent Cells: 5 vials, 200 µl each (total of 1 ml)
• pUC19 DNA (0.01 µg/ml): 1 vial, 100 µl
• SOC Medium: 1 bottle, 6 ml

Store Competent Cells at -80°C. Store pUC19 DNA at -20°C. Store SOC Medium at 4°C or at room temperature.

Documents

Manuals & protocols

Recommended products