This product is manufactured in our Grand Island, US facility and is intended for sale in North America, Latin America, and the Asia/Pacific region—higher freight charges will apply to orders shipped elsewhere. An equivalent product, manufactured in Paisley, Scotland, is recommended for our customers in Europe, Africa, and the Middle East. Click to view.
Gibco™

DMEM, high glucose, pyruvate, no glutamine This product, and/or its packaging, incorporates one or more environmental/green features. Click on this icon to learn more.

Catalog number:  10313021

 Related applications: Mammalian Cell Culture

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

DMEM (Dulbecco's Modified Eagle Medium) is a widely used basal medium for supporting the growth of many different mammalian cells. Cells successfully cultured in DMEM include primary fibroblasts, neurons, glial cells, HUVECs, and smooth muscle cells, as well as cell lines such as HeLa, 293, Cos-7, and PC-12. We offer a variety of DMEM modifications for a range of cell culture applications. Find the right formulation using the media selector tool.

This DMEM is modified as follows:
WithWithout
• High Glucose• L-glutamine
• Phenol Red• HEPES
• Sodium Pyruvate

The complete formulation is available.

Using DMEM
DMEM is unique from other media as it contains 4 times the concentration of amino acids and vitamins than the original Eagle's Minimal Essential Medium. DMEM was originally formulated with low glucose (1 g/L) and sodium pyruvate, but is often used with higher glucose levels, with or without sodium pyruvate. DMEM contains no proteins, lipids, or growth factors. Therefore, DMEM requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). DMEM uses a sodium bicarbonate buffer system (3.7 g/L), and therefore requires a 5–10% CO2 environment to maintain physiological pH.

Product use
For human ex vivo tissue and cell culture processing applications. CAUTION: When used as a medical device, Federal law restricts this device to sale by or on the order of a physician. Customers using Gibco® DMEM in a manufacturing process, who have a submission with the FDA, may request a letter of authorization from us to reference our Type II Drug Master File (DMF).

cGMP manufacturing and quality system
DMEM is manufactured at a cGMP-compliant facility located in Grand Island, New York. The facility is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards. For supply chain continuity, we offer a comparable DMEM product made in our Scotland facility (21969-035). This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard.
For In Vitro Diagnostic Use.

Specifications

Glutamine: No Glutamine
Phenol Red Indicator: Phenol Red
Form: Liquid
Glucose: High Glucose
Serum Supplementation: Standard Serum Supplementation
HEPES Buffer: No HEPES
Sodium Pyruvate Additive: Sodium Pyruvate
Product Line: Gibco®
Product Size: 500 mL
Volume (Metric): 500 ml
Green Features: Sustainable packaging
Shipping Condition: Room Temperature

Contents & storage

Storage conditions: 2-8° C. Protect from light
Shipping conditions: Ambient
Shelf life: 12 months from date of manufacture

Documents

Manuals & protocols

Recommended products