Interested in a bulk or custom version of this product? Request pricing.

FCGR2A (CD32a) Recombinant Human Protein, hIgG1-Fc Tag

Catalog number: 10374H02H5

 Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Fc gamma receptor II-a (FCGR2A/CD32a), recombinant human protein is supplied as a lyophilized powder. In general, recombinant proteins can be used as protein standard and in cell biology research applications.

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Ile 218) of human FCGR2A (AAA35827.1) fused to the Fc region of human IgG1 at the C-terminus.

Activity: This recombinant protein has not been tested.

Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80.

Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2 mg/ml or lower. It is recommended that the protein be aliquoted and be used as soon as possible. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.

Expiration Date: Expires one year from date of receipt when stored as instructed.



This protein is manufactured by Sino Biological Inc.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Protein Family: CD Proteins
Research Category: Immunology, Inflammation, Signal Transduction
Endotoxin Level: < 1 EU/µg
Expression: Mammalian
Form: Lyophilized
Gene Aliases: RP11-5K23.6, CD32, CD32A, CDw32, FCG2, FCGR2, FCGR2A, FCGR2A1, IGFR2, MGC23887, MGC30032
Gene ID: 2212
Gene Symbol: FCGR2A
Molecular Weight: 47.3 kDa
Product Line: Sino Biological®
Product Size: 5 µg
Protein Form: Cell Surface Protein, Recombinant, Truncated
Protein Name: Fc fragment of IgG, low affinity IIa, receptor (CD32)
Protein Subtype: Fc Receptors, Glycoproteins
Protein Tag or Fusion: IgG1 Fc Tag
Purity: ≥ 85% by SDS-PAGE
Species: Human
Validated Application: Assay Standard
Shipping Condition: Room Temperature

Contents & storage

Store at -20 to -70 °C

Documents

Manuals & protocols