Fetal Bovine Serum, qualified, USDA-approved regions This product, and/or its packaging, incorporates one or more environmental/green features. Click on this icon to learn more.

Catalog number:  10437010

Gibco™  Related applications: Mammalian Cell Culture

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Gibco™ fetal bovine sera offers excellent value for basic cell culture, specialty research, and specific assays, earning the trust of researchers with consistent quality and award-winning support that helps meet your research needs and budget requirements

Gibco® Sera: Trusted Quality for Consistent Performance

Sera Category: Standard
• Sera for cell culture with robust cell lines
• Good value for basic research
• Endotoxin level: ≤ 50 EU/ml (levels routinely ≤ 10 EU/ml)
• Hemoglobin level: ≤ 25 mg/dl
• Origin: Countries meeting USDA importation requirements

Gibco® Serum Delivers
• ISO 13485 certified, processed in FDA registered facilities
• Labeled for in-vitro diagnostics (compliant with highest level of USP sterility testing)
• Triple filtered at 0.1 µm
• Gibco® bottle is easier to use in the hood, reduces the risk of contamination and helps you perform cell culture more consistently
• Peel-off sticker on label provides handy reference for lot number and expiration date recording in lab notebook

Specifications

Serum Treatment: Standard (Sterile-Filtered)
Purity or Quality Grade: Qualified
Country of Origin: USDA-Approved
Format: Bottle(s)
Age: Fetal
Product Line: Gibco®
Product Size: 100 mL
Species: Cattle⁄Bovine
Green Features: Sustainable packaging
Shipping Condition: Dry Ice

Contents & storage

Storage conditions: -5 to -20°C
Shipping conditions: Frozen

Documents

Manuals & protocols

Recommended products