Elongase® Enzyme Mix

Catalog number: 10480010

Invitrogen™  Related applications: PCR

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The Elongase® Amplification System is ideal for amplifying DNA fragments up to 30 kb from purified DNA or crude samples (1). The enzyme mix is an optimized mixture of recombinant thermostable DNA polymerases, Taq DNA polymerase, and the proofreading (3'-5' exonuclease activity) enzyme Pyrococcus species GB-D polymerase (Figure 1). Elongase® Amplification System and Enzyme Mix provides:

• Five-times higher fidelity than Taq DNA polymerase alone
• A buffer system that allows amplification of simple and complex genomic targets
• Amplification of long PCR templates or cDNA templates in RT-PCR applications (2)
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Polymerase: Elongase® Enzyme
Hot Start: No
Fidelity (vs. Taq): 5 X
GC-Rich PCR Performance: Low
Reaction Format: SuperMix or Master Mix
Product Overhang: Mixed
Reaction Speed: Standard
Final Product Size(s): 30 kb or less
Product Size: 100 reactions
Exonuclease Activity: 5' - 3', 3' - 5'
Shipping Condition: Dry Ice

Contents & storage

Elongase® Enzyme Mix is supplied with 5X buffer A and 5X buffer B. PCR SuperMix High Fidelity is supplied in a 1.1X concentration. It contains Elongase® Enzyme Mix, dNTPs, salts, and MgSO4. Store at -20°C. Guaranteed stable for 6 months when properly stored.

Documents

Manuals & protocols

Recommended products