Invitrogen™

Terminal Deoxynucleotidyl Transferase, recombinant

Catalog number:  10533073

 Related applications: Restriction Enzyme Cloning

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Terminal Deoxynucleotidyl Transferase, Recombinant (rTdT) is a DNA polymerase that catalyzes the addition of deoxynucleotides to the 3fi hydroxyl terminus of DNA. A TdT Technical Bulletin is available.

Applications:
Homopolymer tailing of vector and insert for cloning. Labeling oligonucleotides with biotin (1,2), 32P- or 35S-label (3), or in apoptosis (TUNEL) (4,5).

Source:
Purified from E. coli clone of calf thymus TdT.

Performance and Quality Testing:
Endonuclease, 3´ and 5´ exodeoxyribonuclease, and levels of incorporation tested.

Unit Definition:
One unit incorporates 1 nmol dATP into acid-precipitable material in 1 h at 37°C, using d(pA)50 as a primer.

Hazard Warning:
Toxic; potassium cacodylate contained in reaction buffer. Also contains cobalt chloride, a highly toxic chemical.
See MSDS.

Unit Reaction Conditions:
0.2 M potassium cacodylate (pH 7.2), 10 mM MgO4 C4 H6 , 1 mM 2-mercaptoethanol, 0.5 mg/ml BSA,
100 flM d(pA)50, 1 mM [3H]dATP, and enzyme in 0.15 ml for 1 h at 37°C.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Enzyme: Terminal Deoxynucleotidyl Transferase
Product Size: 3 x 500 units

Contents & storage

Terminal Deoxynucleotidyl Transferase, Recombinant (rTdT) is supplied with vial of 5X buffer [500 mM potassium cacodylate (pH 7.2), 10 mM CoCl2 , 1 mM DTT]. Store at -20°C.

Documents

Manuals & protocols

Recommended products