Exosome-Human EpCAM Flow Detection Reagent (from cell culture)

Catalog number: 10624D

Invitrogen™  Related applications: Exosomes

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Exosome-Human EpCAM Flow Detection Reagent (from cell culture) enables detection of CD9-positive extracellular vesicles (ECVs) such as exosomes in enriched cell culture media using flow cytometry or electron microscopy. Free exosomes alone are too small to be detected by most flow cytometry instruments. By employing Dynabeads® magnetic separation technology, exosomes can be easily visualized by flow cytometry while bound to the bead surface. The 2.7 µm Dynabeads® magnetic beads allow for a clear and defined flow analysis.

• Minimal hands-on time for specific exosome capture and analysis

• Clear and defined flow analysis in less than 1 hour hands-on time

• Study expression of specific exosome subset markers

• Protocol is easily scalable

Recent studies suggest that the exosome/ECV population consists of several subsets, possibly with different functions. Using the immuno-affinity based Dynabeads® magnetic separation technology, these subsets can be selectively captured and studied using flow cytometry and/or electron microscopy. And with Dynabeads® technology you avoid losing material due to the handling and centrifugation typically experienced with other isolation and analysis methods.

The cell culture media must first be enriched for exosomes. This can be done quickly and efficiently using Total Exosome Isolation Reagent (from cell culture media) or traditional ultracentrifugation.

Note: It is important to provide sufficient mixing to ensure that the magnetic beads do not settle in the sample tube. We recommend using a mixer which provides both tilting and rotation for optimal mixing.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Product Line: Dynabeads®
Product Size: 2 mL
Sample Type (Specific): Cell Cultures

Contents & storage

Store in refrigerator (2–8°C).

Documents

Manuals & protocols