1X CD CHO Medium This product, and/or its packaging, incorporates one or more environmental/green features. Click on this icon to learn more.

Catalog number: 10743011

Gibco™  Related applications: Antibody & Cell Culture Production | Mammalian Cell Culture

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

CD CHO medium is a protein-free, serum-free, chemically-defined medium optimized for the growth of Chinese hamster ovary (CHO) cells and expression of recombinant proteins in suspension culture. With a proven track record of quality for more than 10 years, CD CHO medium contains no proteins or peptide components of animal, plant, or synthetic origin, as well as no undefined lysates or hydrolysates.

When to Use CD CHO Medium
• Your GS CHO cells are recommended to be grown in CD CHO medium
• Your cells have been successfully cultured in CD CHO medium in the past and your process is nearing a regulatory filing (DMF available)

When to Consider Another Gibco® Medium

Consider CD FortiCHO™ medium when:
• The cell line you are using is a transfected CHO K1, GS CHO or CHO-S™ cell line
• Maximum batch culture cell densities and protein titers are needed
• You have time to adapt cells into a new medium in an effort to maximize titers
• Cell health at harvest is a priority for downstream processing

Consider CD OptiCHO™ medium when:
• You are using a transfected CHO cell line other than CHO K1, GS CHO or CHO-S™ cell lines
• Optimization of feeding strategies will be part of your base medium selection testing in an effort to maximize productivity
• Your CHO cell line is “finicky," i.e. hard to grow or adapt

Consider CD DG44 medium when:
• You use parental DG44 or DXB11 CHO cells (dhfr-)

Dry Media Formulations
Gibco® liquid media are available in an AGT™ (Advanced Granulation Technology) dry media format that is designed to provide increased consistency and productivity across all stages of production from development to commercial manufacturing. AGT™ media are manufactured animal origin-free with the same complete, serum-free, protein-free, chemically-defined formulation and performance as their liquid media counterparts.

Design Your Own Media Specific to Your Needs
Whether it’s a bench-level experiment or large scale biomanufacturing process, we offer specialized formats, packaging, and quality levels specific to your needs. With Gibco® cGMP Media Custom Services you can put the manufacturing power and years of technical experience of our R&D and Bioproduction Application Specialists to work for you.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Form: Liquid
Format: Bottle(s)
Species: Hamster
Cell Line: CHO
Glutamine: No Glutamine
Serum Level: Serum-Free
Supplements: No Hypoxanthine, No Thymidine
Concentrated: 1 X
Culture Type: Suspension Cell Culture
Product Size: 500 mL
Classification: Animal Origin-Free, Serum-Free, Chemically Defined, Protein-Free
Endotoxin Level: Low
Volume (Metric): 0.5 L
Included Antibiotics: No Antibiotics
Phenol Red Indicator: No Phenol Red
Sodium Pyruvate Additive: Sodium Pyruvate
Sodium Bicarbonate Buffer: Sodium Bicarbonate
Green Features: Fewer resources used, Less waste
Shipping Condition: Room Temperature

Contents & storage

Store in refrigerator (2–8°C) and protect from light.

Documents

Manuals & protocols

Recommended products