LB Broth This product, and/or its packaging, incorporates one or more environmental/green features. Click on this icon to learn more.

Catalog number:  10855001

Invitrogen™  Related applications: Transformation

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

LB Broth (1X) is a ready-to-use liquid growth media used for bacterial culture. Our filter-sterilized LB Broth can be used for nearly all applications that require a liquid bacterial culturing step, such as protein expression, cloning, and expansion.

LB Broth formulation
LB Broth is one of the most referenced bacterial media for cultivating and maintaining Escherichia coli (E. coli) strains. Our LB Broth is based on the Lennox formulation, one of the most commonly used formulations for LB medium. Formulation per one liter: 10 g SELECT Peptone 140, 5 g SELECT Yeast Extract, 5 g sodium chloride.

Ready-to-use in optimized Gibco® media bottles
Each bottle of LB Broth is filter-sterilized and ready-to-use in ergonomic Gibco® media bottles with angled openings for simplified pipetting.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

LoyaltyEligible: SKUEligibleForLoyalty
Final Product: Liquid Medium
Preparation Method: None (Ready-to-Use)
Form: Liquid
Target Organism Class: E. coli
Product Line: Gibco®
Product Size: 1 L
Green Features: Sustainable packaging
Shipping Condition: Room Temperature

Contents & storage

1 liter LB Broth (1X), liquid; store at room temperature

Documents

Manuals & protocols

Recommended products