Informational:Inquire about OEM or Commercial Supply version of this product here.
Informational:For superior PCR performance, upgrade to Invitrogen's new Platinum II Taq Hot-start DNA Polymerase.
Invitrogen™

Platinum™ Taq DNA Polymerase

Catalog number: 10966034
Invitrogen™

Platinum™ Taq DNA Polymerase

Catalog number: 10966034
Catalog Number
10966034
also known as 10966-034
Unit Size
600 reactions
Price (USD)
Price: 740.00
Online price: 680.65
Your price:
Availability
-
Quantity
Catalog Number
10966018
also known as 10966-018
Unit Size
120 reactions
Price (USD)
Price: 167.00
Online price: 150.65
Your price:
Availability
-
Quantity
Catalog Number
10966026
also known as 10966-026
Unit Size
300 reactions
Price (USD)
Price: 398.00
Online price: 358.65
Your price:
Availability
-
Quantity
Catalog Number
10966083
also known as 10966-083
Unit Size
5,000 reactions
Price (USD)
Price: 5,610.00
Online price: 5,065.65
Your price:
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
10966034
also known as 10966-034
600 reactions
Price: 740.00
Online price: 680.65
Your price:
-
10966018
also known as 10966-018
120 reactions
Price: 167.00
Online price: 150.65
Your price:
-
10966026
also known as 10966-026
300 reactions
Price: 398.00
Online price: 358.65
Your price:
-
10966083
also known as 10966-083
5,000 reactions
Price: 5,610.00
Online price: 5,065.65
Your price:
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Invitrogen Platinum Taq DNA Polymerase is a convenient and reliable 'hot start' thermostable DNA polymerase for PCR that provides enhanced specificity over that of Taq DNA Polymerase. The 'hot start' property of the enzyme is conferred by thermolabile monoclonal antibodies that render Taq DNA polymerase inactive until the initial PCR denaturation step, thus preventing the extention of nonspecifically annealed primers and improving product yield.

Using Platinum Taq DNA Polymerase
The hot-start property of Platinum Taq DNA Polymerase allows for convenient reaction assembly at room temperature. Just as with Taq DNA Polymerase, Platinum Taq DNA Polymerase has a non-template–dependent terminal transferase activity that adds a 3′ deoxyadenosine to product ends, and has a 5′→3′ exonuclease activity. PCR products generated with Platinum Taq DNA Polymerase may be used in the same downstream applications without protocol modifications. Use Platinum Taq DNA Polymerase for the amplification of DNA from complex genomic, viral, and plasmid templates, as well as in RT-PCR.

Note: For superior PCR performance, we recommend next-generation enzyme Platinum II Taq Hot-start DNA Polymerase. Platinum II Taq Hot-Start DNA Polymerase is designed for universal primer annealing and fast, easy PCR with its unique combination of innovative buffer, high-performance engineered Taq DNA polymerase, and superior hot-start technology.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Detection Method
Primer-probe
Format
Tube
GC-Rich PCR Performance
Low
Polymerase
Platinum Taq DNA Polymerase
Reaction Speed
Fast or Standard
Exonuclease Activity
5' - 3'
Product Line
Platinum™
Product Type
DNA Polymerase
Quantity
600 Reactions
Shipping Condition
Approved for shipment on Wet or Dry Ice
Sufficient For
600 Reactions
Fidelity (vs. Taq)
1 X
Hot Start
Built-In Hot Start
No. of Reactions
600 Reactions
Overhang
3'-A
Reaction Format
Separate Components
Size (Final Product)
5 kb or less
Starting Material
DNA

Contents & Storage

• 1 x 120 μL Platinum Taq DNA Polymerase
• 3 x 1.25 mL 10X PCR Buffer (no magnesium chloride)
• 1 x 1 mL 50 mM MgCl2
• 1 x 1.3 mL KB Extender

Store at -10 to -30°C in a non-frost-free freezer.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text