Interested in a bulk or custom version of this product? Request pricing.

TREM2 Recombinant Human Protein, His Tag

Catalog number: 11084H08H50

 Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Manufactured by Sino Biological (China); Distributed by Life Technologies.

Triggering Receptor Expressed On Myeloid Cells 2 (TREM2) recombinant human protein is supplied as a lyophilized powder. This protein is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

This recombinant human protein is expressed from a DNA sequence encoding the extracellular domain of human TREM2 (NP_061838.1) (Met 1-Ser 174) fused to a polyhistidine tag at the C-terminus.

N-terminal Sequence Analysis: His 19.

Activity: The activity of this protein has not been tested.

Formulation: lyophilized in PBS, pH7.4, 5% mannitol, 5% trehalose, 0.02% Tween®-80.

Reconstitution: dissolve the protein in sterile double-distilled water to a concentration of 0.2 mg/mL or lower. It is recommended that the protein be aliquoted and used as soon as possible. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.

Expiration Date: expires one year from date of receipt when stored as instructed.

Background
Triggering receptor expressed on myeloid cells 2 ( TREM2 ) is a single Ig domain receptor. It is expressed on macrophages and dendritic cells but not on granulocytes or monocytes. TREM2 may play a role in chronic inflammations and may stimulate production of constitutive rather than inflammatory chemokines and cytokines. TREM2 forms a receptor signaling complex with TYROBP and triggers activation of the immune responses in macrophages and dendritic cells. It also associates with the signal adapter protein, DAP12, which has a cytoplasmic ITAM, leading to the subsequent activation of cytoplasmic tyrosine kinases. TREM2 is both required and sufficient for competent uptake of apoptotic neuronal cells. TREM2 and TREM2-L form a receptor-ligand pair connecting microglia with apoptotic neurons, directing removal of damaged cells to allow repair. Deficiency of the adapter protein DAP12 or its associated receptor TREM2 is associated with abnormal osteoclast development in humans. Defects in TREM2 are causes of PLOSL, also known as NHD. In addition, TREM2 signaling is also an important pathway to promote healing.

You may also be interested in the following TREM2 products:
Enzyme & Protein Assays
Gene Expression Assays
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Protein Family: Other Proteins
Research Category: Immunology, Signal Transduction
Endotoxin Level: < 1 EU/µg
Expression: Mammalian
Form: Lyophilized
Gene Aliases: TREM-2, Trem2a, Trem2b, Trem2c, TREM2
Gene ID: 54209
Gene Symbol: TREM2
Molecular Weight: 18.9 kDa
Product Line: Sino Biological®
Product Size: 50 µg
Protein Form: Recombinant, Truncated
Protein Name: triggering receptor expressed on myeloid cells 2
Protein Tag or Fusion: His Tag
Purification: PAGE
Purity: ≥ 85% by SDS-PAGE
Species: Human
Tag Position: C-Terminal
Validated Application: Assay Standard

Contents & storage

Includes 1 vials of protein in suspension buffer. Store at -20 °C..

Documents

Manuals & protocols