Related applications: Cell Isolation & Expansion
Error loading your content!
Catalog number |
Select a plan
Unit size
|
Price ({{currency}}) | Availability | Qty | |
---|---|---|---|---|---|
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} |
|
— |
Capacity: | Processes ~2x10^9 cells total |
---|---|
Cell Type: | Monocytes |
Isolation Method: | Depletion, positive isolation for molecular applications |
Output Viability: | >95% |
Product Line: | DYNAL®, Dynabeads® |
Product Size: | 5 mL |
Sample Type (Specific): | Blood, PBMC |
Starting Material (Cell #): | 1x10^7 PBMCs per isolation |
Target Species: | Human |
Shipping Condition: | Room Temperature |
Catalog number: | 11350D |
Size | 1 kit |
Price | USD 738.00 |
Add to favorites | |
Catalog number: | 11367D |
Size | 2 x 3 mL |
Price | USD 1,110.00 |
Add to favorites | |
Catalog number: | C34557 |
Size | 180 assays |
Price | USD 379.00 |
Add to favorites | |