Invitrogen™

Dynabeads™ CD45

Catalog number:  11153D

 Related applications: Cell Isolation & Expansion

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Dynabeads® CD45 are superparamagnetic beads covalently coupled to anti-human CD45 antibody that enable isolation or depletion of CD45+ leucocytes directly from whole blood, buffy coat, or MNC suspensions. Dynabeads® CD45 can also be used to enrich epithelial tumor cells. The CD45 antigen is weakly expressed on myeloid cells. To efficiently deplete or isolate all leucocytes including myeloid cells, use Dynabeads® CD15 (Cat. No. 11137D) in combination with Dynabeads® CD45. Advantages of Dynabeads® CD45:

• Efficient depletion of human leucocytes
• Isolation of leucocytes directly from whole blood for molecular applications
• Enrichment of circulating epithelial tumor cells

About Dynabeads® CD45
Dynabeads® CD45 are uniform, superparamagnetic beads (4.5 µm diameter) coated with a primary monoclonal mouse IgG2a antibody specific for a CD45 membrane antigen common to all known isoforms of CD45. CD45 is expressed on all human leucocytes. Because of the bead size, Dynabeads® CD45 can easily and efficiently isolate or deplete cells from viscous samples such as whole blood and bone marrow in about 30 minutes. Dynabeads® CD45 is often used for enrichment of circulating non-hematopoietic tumor cells, by depletion of all CD45+ leucecytes from a MNC sample. Positively isolated cells can be used for downstream molecular studies; for example, those in which cells are to be lysed while still attached to beads, and nucleic acids or proteins further purified. Note that intact cells will not be released from these beads, and we do not recommend to analyze the bead-bound cells with a flow cytometer.

Magnetic bead–based separation offers easy handling
Dynabeads® CD45 is added to the sample under continuous mixing to optimize the binding of the Dynabeads® to the target cells. By placing the sample on a magnet it separates the bead-bound target cells from the rest of the sample in just 1–2 minutes. For depletion, remove the supernatant to a new tube for further studies and discard the bead-bound cells. For positive isolation for molecular studies, remove the supernatant and wash the bead-bound cells 2–3 times in buffer to get optimal purity. The cells can be lysed while still attached to the beads and the supernatant transferred to a new tube for downstream molecular analysis. Starting samples can be whole blood, buffy coat, bone marrow, PBMC, or tissue digests.

Learn more about Dynabeads® products
• Find Dynabeads® products for a whole range of applications.
• Find magnets for Dynabeads® separations.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Capacity: Processes ~5x10^8 cells total
Cell Type: Leukocytes, Tumor Cells
Isolation Method: Depletion, positive isolation for molecular applications
Output Viability: >95%
Product Line: DYNAL®, Dynabeads®
Product Size: 5 mL
Sample Type (Specific): Blood, PBMC
Starting Material (Cell #): 1x10^7 PBMCs per isolation
Target Species: Human
Shipping Condition: Room Temperature

Contents & storage

This Product Contains: 5 ml Dynabeads® CD45 coated with anti-CD45 monoclonal antibody.

Storage Conditions: 2°C to 8°C. Do not freeze.

Documents

Manuals & protocols

Recommended products