Dynabeads® Human T-Activator CD3/CD28/CD137

Catalog number: 11163D

Gibco™  Related applications: Cell Isolation & Expansion

Error loading your content!

  Catalog number
Select a plan
Unit size
List price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Dynabeads® Human T-Activator are coupled with anti-CD3, anti-CD28, and anti-CD137 antibodies. CD137 (4-1BB) is a member of the tumor necrosis factor family. Agonistic anti-CD137 antibody acts as an activating costimulatory molecule especially important for effector/memory T cells and promotes the survival and proliferation of T lymphocytes.

• Efficient expansion and increased survival rate of Ag-specific T cells
• No need to find or prepare donor-matched MNC or antigen
• T cells expanded with Dynabeads® maintain Ag-specific responses
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Capacity: 100 tests
Cell Type: T Cells (Activated)
Reactivity: Human
Product Size: 2 mL
Final Product: Cells
Target Species: Human
Shipping Condition: Room Temperature

Contents & storage

2 ml Dynabeads® Human T-Activator CD3/CD28/CD137 is supplied as a suspension containing 5 x 105 Dynabeads®/ml in phosphate buffered saline (PBS), pH 7.4, with 0.1% human serum albumin (HSA). Store at: 2-8°C.

Documents

Manuals & protocols