Dynabeads® M-280 Streptavidin

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Dynabeads® M-280 Streptavidin are the gold standard for the isolation and handling of biotinylated nucleic acids, antibodies, or other biotinylated ligands and targets. The strong binding affinity of the streptavidin-biotin interaction (Kd=10-15) is used in a vast number of applications.

Benefits and features include:

• Direct and fast isolation of any biotinylated molecule
• Low-charged and neutral beads, optimal for binding of DNA fragments, proteins, peptides, and antibodies
• Flexible protocols with gentle and efficient liquid-phase reaction kinetics
• Biomagnetic protocols are easily adapted to automated platforms
• High batch-to-batch reproducibility, securing consistent results in your application

About Dynabeads® M-280 Streptavidin
These uniform and superparamagnetic beads are 2.8 µm in diameter, with a monolayer, not a multilayer, of recombinant streptavidin covalently coupled to the surface and further blocked with BSA. The monolayer of streptavidin leaves the vast majority of the biotin binding sites sterically available for binding, not only of free biotin, but also for binding of biotinylated ligands/targets. They show rapid liquid-phase reaction kinetics. Their specific and defined surface allow for efficient capture, separation, and downstream handling. The streptavidin monolayer ensures negligible leakage, and the lack of excess adsorbed streptavidin ensures batch consistency and reproducibility of your results.

Applications
Over the past 15 years, Dynabeads® M-280 Streptavidin have been used and cited for a very wide variety of applications. Key applications include preparing 2–5 kb samples for mate pair library sequencing using Illumina platforms preparing single-stranded DNA templates, isolation of RNA and DNA binding proteins, immobilization of large DNA fragments, purifying sequencing products, and the specific capture of nucleic acids. Dynabeads® are used on more than 25,000 routine IVD instruments worldwide. The product holds high standards with respect to reproducibility (both within and between batches), and automation ability, and drives reliability for your results.

Binding capacity
The size of the molecule and the biotinylation procedure will affect the binding capacity. The capacity also depends on steric availability and charge interaction between bead and molecule and between molecules. There are two or three biotin binding sites available for each streptavidin molecule on the surface of the bead after immobilization. One mg of Dynabeads® M-280 Streptavidin typically binds:

• 650–900 pmoles free biotin
• ~200 pmol biotinylated peptides
• ~10 µg biotinylated IgG
• ~10 µg ds-DNA
• ~200 pmol ss-oligonucleotides
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Ligand Type: Streptavidin
Form: Beads in Suspension
Shelf Life: 36 months from date of manufacture
Product Size: 2 mL
Bead Diameter: 2.8 µm
Concentration: 10 mg⁄ml
Binding Property: 650-900 pmoles/mg beads
Isoelectric Point: pH 5.0
Size Distribution: CV < 3%
Surface Functionality: Hydrophobic, Tosylactivated
Sample Type (Specific): Any Sample Type
Iron Content (Ferrites): 12%
High Throughput Compatibility: High Throughput-Compatible
Shipping Condition: Room Temperature

Contents & storage

This Product Contains: Dynabeads® (2.8 µm) with covalently coupled recombinant streptavidin. Supplied in PBS, pH 7.4 with 0.1% BSA and 0.02% sodium azide.

Store at: 2°C to 8°C.

Documents

Manuals & protocols

Recommended products