This product is manufactured in our Grand Island, US facility and is intended for sale in North America, Latin America, and the Asia/Pacific region—higher freight charges will apply to orders shipped elsewhere. An equivalent product, manufactured in Paisley, Scotland, is recommended for our customers in Europe, Africa, and the Middle East. Click to view.

Waymouth's Medium This product, and/or its packaging, incorporates one or more environmental/green features. Click on this icon to learn more.

Catalog number:  11220035

Gibco™  Related applications: Mammalian Cell Culture

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top


Waymouth's is a chemically defined medium developed by Charity Waymouth and originally formulated for studies on the nutrition, metabolism, and growth of mouse Strain L sublines, NCTC clone 929 (L929 cells) in a serum-free environment. Waymouth's medium is also used for the culture of whole organs and a broad range of cell types, such as carcinoma cell lines.
This Waymouth's medium is modified as follows:
• L-glutamine• HEPES
• Phenol Red

The complete formulation is available.

Product Intended Use
For in vitro diagnostic use. CAUTION: Not for human or animal therapeutic use. Uses other than the labeled intended use may be a violation of local law.

Dual-Site cGMP Manufacturing
Gibco® Waymouth's medium is manufactured at a cGMP compliant facility, located in Grand Island, New York. The facility is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards. For supply chain continuity, we offer an identical Gibco® Waymouth's product made in our Scotland facility (31220-023). This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. Waymouth's contains no proteins, lipids, or growth factors. Some cell types may required the supplementation with Fetal Bovine Serum (FBS). Waymouth's uses a sodium bicarbonate buffer system (2.24 g/L) and therefore requires a 5-10% CO2 environment to maintain physiological pH.
For In Vitro Diagnostic Use.


Form: Liquid
Glucose: High Glucose
Glutamine: L-Glutamine
Phenol Red Indicator: Phenol Red
Product Line: Gibco®
Product Size: 500 mL
Sodium Pyruvate Additive: No Sodium Pyruvate
Volume (Metric): 500 ml
Green Features: Sustainable packaging
Shipping Condition: Room Temperature

Contents & storage

Storage conditions: 2-8°C. Protect from light
Shipping conditions: Ambient
Shelf life: 12 months from date of manufacture

Media formulations


Manuals & protocols

Recommended products