Dynabeads™ Human DC Enrichment Kit

Catalog number:  11308D

Invitrogen™  Related applications: Cell Isolation & Expansion

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Use this kit to enrich viable human dendritic cells (DC's) from PBMC by negative isolation. The kit depletes T cells, B cells, monocytes/ macrophages, granulocytes, NK cells and erythrocytes. Highly enriched DC's are bead and antibody-free, and can be used in any functional assay. The cells should be further sorted to obtain high purity of the specific DC sub-populations of choice.

• Obtain high recovery (80-100%) of viable DC's of all sup-populations.
• Substantial reduction in sorting time and volume compared to sorting only, thus saving money as well.

Methodology:
An antibody mix towards the non- dendritic cells is added to the sample and allowed to bind to the cells. Dynabeads® are added and will bind to the antibody-labeled cells during a short incubation. The bead-bound cells are quickly separated on a magnet and discarded. The remaining untouched human DC's can be directly sorted into the sup-population of choice or used in any other application.

Starting samples:
PBMC
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Capacity: Processes ~2x10^9 cells total
Cell Type: Dendritic Cells
Isolation Method: Negative Isolation
Output Viability: >95%
Product Line: DYNAL®, Dynabeads®
Product Size: 2 x 10 mL
Sample Type (Specific): PBMC
Starting Material (Cell #): 1x10^7 PBMCs per isolation
Target Species: Human
Shipping Condition: Room Temperature

Contents & storage

The Kit Contains: 2x10 ml Depletion MyOne™ Streptavidin Dynabeads® and 4 ml Antibody Mix containing an optimized mixture of biotinylated monoclonal antibodies towards CD3, CD14, CD16, CD19, CD56 and Glycophorin A.
Store at 2-8°C

Documents

Manuals & protocols

Recommended products