Dynabeads® Untouched™ Human NK Cells Kit

Catalog number: 11349D

Invitrogen™  Related applications: Cell Isolation & Expansion

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Use this kit to isolate pure and viable untouched NK cells from PBMC by negative isolation. The kit depletes T cells, B cells, monocytes, platelets, dendritic cells, granulocytes and erythrocytes. The negatively isolated human NK cells are left in the sample and have not been in contact with the Dynabeads. The bead and antibody-free NK cells are truly untouched, and in perfect shape for any functional assay and application.

• High purity, recovery and viability of human untouched NK cells
• Easy to use and to scale up
• No columns required

Methodology:
An antibody mix towards the non-NK cells is added to the sample and allowed to bind to the cells. Dynabeads® are added and will bind to the antibody-labeled cells during a short incubation. The bead-bound cells are quickly separated on a magnet and discarded. The remaining negatively isolated and untouched human NK cells can be directly analyzed in a flow cytometer and used in any application.

Starting Sample:
PBMC
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Capacity: Processes ~1x10^9 cells⁄kit
Cell Type: NK Cells
Final Product: Cells
Form: Bead
Isolation Method: Negative Isolation
Number of Reactions: 20 Reactions
Output Viability: >99%
Product Line: DYNAL®, Dynabeads®, Untouched™
Product Size: 1 kit
Sample Type (Specific): MNC
Starting Material (Cell #): ~5x10^7 cells/test
Target Species: Human
Shipping Condition: Room Temperature

Contents & storage

The Kit Contains: 10 ml Depletion MyOne™ SA Dynabeads® and 2 ml Antibody Mix with biotinylated monoclonal antibodies towards human CD3, CD14, CD36, HLA Class II, CD123 and CD235a (Glycophorin A).
Store at 2-8°C

Documents

Manuals & protocols