Dynabeads® FlowComp™ Human CD8 Kit

Catalog number: 11362D

Invitrogen™  Related applications: Cell Isolation & Expansion

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

This gentle and easy-to-use kit allows you to isolate flow-compatible human CD8+ T-cells from PBMC or directly from whole blood. Immediately after positive isolation, the beads are released and removed from your cell sample. In this way, you avoid exposing your cells to potentially cytotoxic or immunogenic foreign substances like iron oxides or dextrans, a potential problem when working with biodegradable particles. In addition, your cells will not be exposed to the stress of being passed through a column. You can expect a purity level of >95% human CD8+ cells.

Advantages:

• No risk of your results being affected by the isolation method
• Gentle tube-based method - no columns needed
• High purity and recovery
• Bead-free cells - compatible with flow cytometry analysis
• Compatible with flow cytometry analysis
• Viable, bead-free CD8+ T-cells for direct use in any cell-based assay.

The isolated cells have the presumed profile of memory and naive CD8+ T cells, and show a high proliferation capacity. You will save time and money, and get increased confidence in your results.

Starting sample:
MNC or whole blood.

Capacity:
The kit will process 2 × 109 mononuclear cells (MNC) or 80 ml whole blood
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Capacity: Processes ~2x10^9 cells total
Cell Type: T Cells (CD8)
Product Size: 3 mL
Target Species: Human
Isolation Method: Positive Isolation
Output Viability: >95%
Sample Type (Specific): Blood, PBMC
Starting Material (Cell #): 5x10^7 PBMCs or 2 ml whole blood per isolation
Shipping Condition: Room Temperature

Contents & storage

The Kit Contains 1 ml Antibody, 3 ml FlowComp™ Dynabeads® and 40 ml FlowComp™ Release Buffer.
Store at: 2-8°C

Documents

Manuals & protocols

Recommended products