Dynabeads® Mouse CD43 (Untouched™ B Cells)

Catalog number: 11422D

Invitrogen™  Related applications: Cell Isolation & Expansion

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Use this kit to isolate pure and viable untouched B cells from mouse lymphoid organs by negative isolation. The kit depletes T cells, monocytes/macrophages, NK cells, plasma cells, CD5+ B1 cells, platelets, dendritic cells, granulocytes and erythrocytes. The negatively isolated mouse B cells are left in the sample and have not been in contact with the Dynabeads. The bead and antibody-free B cells are truly untouched, and in perfect shape for any functional assay and application.

High purity, recovery and viability of mouse untouched B cells

• Easy to use and to scale up
• No columns required

Methodology:
Dynabeads Mouse CD43 (Untouched B Cells) is added directly to the sample and binds the unwanted cells. The bead-bound cells are quickly separated on a magnet and discarded. The remaining negatively isolated and untouched mouse B cells can be directly analyzed in a flow cytometer and used in any application.

Starting Sample:
spleen or lymph nodes
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Form: Bead
Capacity: Processes ~2x10^9 cells total, Processes ~2x10^9 cells⁄kit
Cell Type: B Cells
Reactivity: Mouse
Product Size: 1 kit
Final Product: Cells
Target Species: Mouse
Isolation Method: Negative Isolation
Number of Reactions: 40 Tests
Sample Type (Specific): Spleen, Lymph Node, Tissue Digests
Starting Material (Cell #): 5x10^7 cells⁄test
Shipping Condition: Room Temperature

Contents & storage

The Kit Contains: 5 ml MyOne™ Dynabeads®- coated with streptavidin and biotinylated rat monoclonal CD43 antibody
against mouse T cells, monocytes⁄macrophages, NK cells, plasma cells, CD5+ B1 cells, platelets, dendritic cells, granulocytes and erythrocytes.
Store at 2-8°C

Documents

Manuals & protocols

Recommended products