Dynabeads™ Mouse CD8 (Lyt 2)

Catalog number:  11447D

Invitrogen™  Related applications: Cell Isolation & Expansion

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Dynabeads® Mouse CD8 (Lyt 2) are magnetic beads that provide a robust and versatile tool to isolate or deplete mouse CD8+ T-cells directly from any sample, including tissue digests such as spleen or lymph nodes, whole blood, bone marrow, and mononuclear cells (MNC).
• Rapid isolation of mouse CD8+ T-cells from any sample – no columns required
• Positive isolation for molecular downstream assays or efficient depletion of mouse CD8+ T cells

Excellent Yield, Purity, and Viability of Recovered Cells
Dynabeads® Mouse CD8 (Lyt 2) are uniform, superparamagnetic beads (4.5 µm diameter) that enable easy isolation of mouse CD8+ cells from spleen or lymph nodes, although any single-cell suspension may be used. Dynabeads® Mouse CD8 beads are coated with a primary monoclonal antibody specific for the Lyt 2 membrane antigen mainly expressed on cytotoxic/suppressor subpopulation of mature T-cells of all common mouse strains. Dynabeads® Mouse CD8 binds to CD8+ cells during a short incubation period in a tube, after which the bead-bound cells are separated from unbound cells using a magnet. This fast and gentle isolation method does not require the use of columns, and helps ensure high purity, recovery and viability of the isolated CD8+ cells.

Ideal for Both Depletion and Positive Isolation for Downstream Molecular Assays
Because of the powerful magnets used for separation, Dynabeads® Mouse CD8 (Lyt 2) can be used in viscous samples such as single cell suspensions from tissue, whole blood and bone marrow, and enable efficient depletion of mouse CD8+ cells in about 30 minutes. Dynabeads® Mouse CD8 (Lyt 2) will also afford excellent recovery of high-purity, viable cells when used for positive isolation for downstream studies; for example, those in which cells are to be lysed while still attached to beads, and nucleic acids or proteins further purified. Note that intact cells will not be released from these beads, so if you want to isolate mouse CD8+ cells for cell-based applications, or you need to check your samples with flow cytometry, you should use Dynabeads® FlowComp Mouse CD8 Cells (for bead-free cells).

Select what is right for you from our range of Mouse CD8 Isolation Products.

For Research Use Only. Not for human or animal therapeutic or diagnostic use.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Capacity: Processes ~2x10^9 cells total
Cell Type: T Cells (CD8)
Isolation Method: Depletion, positive isolation for molecular applications
Output Viability: >95%
Product Line: DYNAL®, Dynabeads®
Product Size: 5 mL
Reactivity: Mouse
Sample Type (Specific): Blood, Lymph Node, Spleen
Starting Material (Cell #): 1x10^7 cells per isolation
Target Species: Mouse
Shipping Condition: Room Temperature

Contents & storage

This Product Contains: 5 ml Dynabeads® Mouse CD8 (Ly-2), coated with an anti-Ly2 monoclonal antibody.
Lyt-2 antigen is expressed on thymocytes and the cytotoxic/ suppressor subset of
mature T cells from all common mouse strains.
Store at 2-8°C

Documents

Manuals & protocols

Recommended products