Taq PCRx DNA Polymerase

Catalog number: 11508017

Invitrogen™  Related applications: PCR

Error loading your content!

  Catalog number
Select a plan
Unit size
List price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Taq PCRx DNA Polymerase is recombinant Taq DNA polymerase supplied with the PCRx Enhancer System for optimizing PCR of problematic and/or GC-rich templates. With Taq PCRx DNA Polymerase you'll get:

• Higher primer specificity, broader magnesium concentration, broader annealing temperature, and improved thermostabilty of Taq DNA polymerase
• Amplification of PCR products up to 5 kb in size
• A system that is licensed and qualified for PCR

Applications: Amplification of DNA from complex genomic, viral, and plasmid templates; and RT-PCR. Especially effective for problematic tem-plates such as GC-rich DNA.
Unit Definition: One unit incorporates 10 nmol of deoxyribonucleotide into DNA in 30 min at 74°C.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Polymerase: Taq DNA Polymerase
Hot Start: No
Fidelity (vs. Taq): 1 X
GC-Rich PCR Performance: High
Reaction Format: Separate Components
Product Overhang: 3'-A
Reaction Speed: Standard
Final Product Size(s): 5 kb or less
Product Size: 500 units
Exonuclease Activity: 5' - 3'
Shipping Condition: Approved for shipment on Wet or Dry Ice

Contents & storage

Taq PCRx DNA Polymerase is supplied with 10X PCR Buffer, 10X PCRx Enhancer Solution, 50 mM MgS0 4 , 10X Amplification Buffer, and 50 mM MgCl 2 . Store at -20°C. Guaranteed stable for 6 months when stored properly.

Documents

Manuals & protocols

Recommended products