Invitrogen™

CloneChecker™ System (for screening bacterial colonies)

Catalog number:  11666013

 Related applications: DNA Extraction

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

CloneChecker™ System is a rapid method for screening recombinant bacterial colonies or liquid cultures of colonies for the presence of target plasmid DNA (1). The CloneChecker™ System:

• Screens bacterial colonies for plasmid DNA and insert of interest prior to purification
• Screens for inserts >200 bp in plasmids up to 15 kb by size comparison to the original vector or a DNA size marker
• Goes from plated colonies through lysis in less than 5 minutes
• Allows for supercoiled, restriction digest, or PCR analysis
• Works with single-copy plasmids starting with a 2-mm bacterial colony
• Includes sufficient reagents for processing 100 bacterial samples by supercoiled analysis (direct size comparison); and 100 samples by restriction endonuclease or PCR analysis
For Research Use Only. Not for use in diagnostic procedures.

Figures

Specifications

High Throughput Compatibility: Not High Throughput-Compatible (Manual)
Sample Type (General): Bacteria
Downstream Application: Cloning, PCR
Final Product: Plasmid DNA
Product Line: CloneChecker™
Product Size: 100 preps
Sample Type (Specific): Bacterial Culture
Shipping Condition: Room Temperature

Contents & storage

The CloneChecker™ System includes Yellow, Red, and Green solutions and Loading buffer. Store at 15°C to 30°C.

Documents

Manuals & protocols

Recommended products