Gibco™

VP-SFM (1X) This product, and/or its packaging, incorporates one or more environmental/green features. Click on this icon to learn more.

Catalog number:  11681020

 Related applications: Mammalian Cell Culture | Viral Vaccine Production

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

VP-SFM is a serum-free, ultra-low protein (5 µg/ml) medium containing no proteins, peptides, or other components of animal or human origin. It is designed for the growth of VERO cells for virus production (1). This medium is suitable for the growth of COS-7, MDCK, BHK-21 (suspension culture) and HEp2 cells, and for recombinant protein production.

Formulated without L-glutamine for increased stability; add 20 ml of 200 mM L-glutamine or 20 ml of GlutaMAX™-I supplement per 1,000 ml of medium prior to use.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Cell Line: BHK-21, COS-7, HEp2, MDCK, VERO
Classification: Animal Origin-Free, Low-Protein, Serum-Free
Concentrated: 1 X
Culture Type: Suspension Cell Culture
Endotoxin Level: Low
Form: Liquid
Format: Bottle(s)
Glutamine: No Glutamine
Included Antibiotics: No antibiotics
Phenol Red Indicator: Phenol Red
Product Line: Gibco®
Product Size: 1,000 mL
Sodium Bicarbonate Buffer: Sodium Bicarbonate
Sodium Pyruvate Additive: Sodium Pyruvate
Volume (Metric): 1 L
Green Features: Sustainable packaging
Shipping Condition: Room Temperature

Contents & storage

Store in refrigerator (2–8°C) and protect from light.

Documents

Manuals & protocols

Recommended products