Interested in a bulk or custom version of this product? Request pricing.

USP7 Recombinant Human Protein

Catalog number: 11681HNCB250

 Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Ubiquitin Carboxyl-Terminal Hydrolase 7 (USP7), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

This recombinant protein was expressed from a DNA sequence encoding the amino acid sequence (Lys 208-Glu 560) of human USP7 (NP_003461.2) fused to two additional aa (Gly & Pro) at the N-terminus.

Activity: The activity of this protein has not been tested.

Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80.

Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2 mg/ml or lower. It is recommended that the protein be aliquoted and be used as soon as possible. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.

This protein is manufactured by Sino Biological Inc.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Protein Family: Proteases Peptidases & Inhibitors
Research Category: Neurobiology, Signal Transduction
Endotoxin Level: < 1 EU/µg
Expression: Insect
Form: Lyophilized
Gene Aliases: HAUSP, TEF1
Gene ID: 7874
Gene Symbol: USP7
Molecular Weight: 41 kDa
Product Line: Sino Biological®
Product Size: 5 x 50 µg
Protein Form: Recombinant, Truncated
Protein Name: Ubiquitin carboxyl-terminal hydrolase 7
Purification: PAGE
Purity: ≥ 95% by SDS-PAGE
Species: Human
Tag Position: None
Validated Application: Assay Standard

Contents & storage

Includes 5 vials of protein in suspension buffer. Store at -20 °C..

Documents

Manuals & protocols