Gibco™

293 SFM II This product, and/or its packaging, incorporates one or more environmental/green features. Click on this icon to learn more.

Catalog number:  11686029

 Related applications: Antibody & Cell Culture Production | Mammalian Cell Culture | Viral Vaccine Production

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

293 SFM II is a serum-free, low-protein (<10 µg/ml) formulation optimized for high-density suspension culture of 293 cells (1). (It is not suitable for adherent 293 cell cultures.) This product replaces 293 SFM (Cat. No. 10970-010). 293 SFM II is animal-origin free. Formulated without L-glutamine for increased stability; add 20 ml of 200 mM L-glutamine or 20 ml of GlutaMAX™-I Supplement per 1,000 ml of medium prior to use.

Note:
A 293 Medium Master file has been submitted to the FDA. Permission to cross-reference the Master File may be obtained by contacting Technical Services or your local Sales Representative.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Cell Line: 293 (HEK), 293F
Classification: Animal Origin-Free, Low-Protein, Serum-Free
Culture Type: Suspension Cell Culture
Form: Liquid
Glutamine: No Glutamine
Product Line: Gibco®
Product Size: 1,000 mL
Serum Level: Serum-Free
Species: Human
Green Features: Sustainable packaging
Shipping Condition: Room Temperature

Contents & storage

Store in refrigerator (2–8°C) and protect from light.

Documents

Manuals & protocols

Recommended products