Upgrade to Invitrogen™ Platinum™ SuperFi™ DNA Polymerase: >100-fold higher fidelity than Taq, robust and versatile.

Platinum™ Pfx DNA Polymerase

Catalog number:  11708021

Invitrogen™  Related applications: Chromatin Biology | Methylation Analysis | PCR

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Platinum® Pfx DNA Polymerase is ideal for amplification of DNA fragments for high-fidelity PCR applications. High fidelity is provided by a proprietary enzyme preparation containing recombinant DNA polymerase from Thermococcus species KOD with proofreading (3´→5´ exonuclease) activity. Platinum® antibody technology provides a simple, automatic, hot-start method that improves PCR specificity (see figure). PCRx Enhancer Solution is included for higher primer specificity, broader magnesium concentration allowances, broader annealing temperature parameters, and improved thermostability of Platinum® Pfx DNA Polymerase. The PCRx Enhancer Solution also helps optimize PCR of problematic and/or GC-rich templates. Platinum® Pfx DNA Polymerase provides:

• 26 times higher fidelity than Taq DNA polymerase
• Amplification of fragments up to 12 kb
• Room temperature reaction assemblyApplications
Amplification of DNA from complex genomic, viral, and plasmid templates; and RT-PCR. Platinum® Pfx DNA Polymerase is especially effective when very high levels of fidelity are required.

Unit definition
One unit incorporates 10 nmoles of deoxyribonucleotide into acid-precipitable material in 30 min at 74°C.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Polymerase: Platinum® Pfx DNA Polymerase
Hot Start: Built-In Hot Start
Fidelity (vs. Taq): 26 X
GC-Rich PCR Performance: Medium
Reaction Format: Separate Components
Product Overhang: Blunt
Reaction Speed: Standard
Final Product Size(s): 12 kb or less
Exonuclease Activity: 3' - 5'
Product Line: Platinum®
Product Size: 250 reactions
Shipping Condition: Approved for shipment on Wet or Dry Ice

Contents & storage

Platinum® Pfx DNA Polymerase (250 units total, at 2.5U/µL) is a supplied with a vial (2 × 1 mL) of 10X PCRx Enhancer solution, a vial (2 × 1 mL) of 10X Pfx amplification buffer, and a vial (1 mL) of 50 mM MgSO4.

Designed for 250 reactions (at 1 unit each in a 50 µL reaction volume). Store at -20°C.

Documents

Manuals & protocols

Recommended products