Platinum® Quantitative PCR SuperMix-UDG

Catalog number: 11730017

Invitrogen™  Related applications: Chromatin Biology | Real Time PCR (qPCR) | Real Time PCR-Based Gene Expression Profiling

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

**********************
Alternative Product: Try TaqMan Fast Advanced Master Mix, our highest-performance, probe-based master mix. With TaqMan Fast Advanced Master Mix, we’ve taken the best of Platinum Quantitative PCR SuperMix and added additional capabilities for your gene expression analysis.

**********************
Platinum® Quantitative PCR SuperMix-UDG is a ready-to-use mixture for high-specificity, real-time quantitative DNA amplification, containing UDG to prevent carryover contamination (1,2). The system is optimized to provide superior performance with either LUX™ Fluorogenic Primers or probe-based detection systems.

Platinum® Quantitative PCR SuperMix-UDG:

• Is highly sensitive and specific
• Allows room temperature reaction assembly (3)
• Is easy to use-just add template and fluorogenic primers or probes for detection
• Includes a vial of 50 mM MgCl2 to accommodate a variety of primer/template PCR systems
• Is compatible with most real-time instruments

Applications: Real-time quantitative amplification of DNA from complex genomic, viral, and plasmid templates, RT-PCR, and fluorometric SNP (Single Nucleotide Polymorphism) detection.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Detection Method: Primer-Probe Detection
For Use With (Equipment): 7500 System, BioRad MyiQ, BioRad iCycler iQ, BioRad iQ5, Cepheid SmartCycler, MJ Chromo4, MJ Opticon, Stratagene Mx3000P, Stratagene Mx3005P, Stratagene Mx4000
Reaction Speed: Standard
PCR Method: qPCR
Sample Type (General): DNA
Product Line: Platinum®
Product Size: 100 reactions
Sample Type (Specific): DNA (Genomic), cDNA
Shipping Condition: Dry Ice

Contents & storage

Platinum® Quantitative PCR SuperMix-UDG is supplied in a 2X concentration [40 mM Tris-HCl (pH 8.4), 100 mM KCl, 6 mM MgCl2 , 400 µM dATP, 400 µM dCTP, 400 µM dGTP, 800 µM dUTP, 40 units/ml UDG, 60 units/ml Platinum® Taq DNA Polymerase, stabilizers]. A vial of 50 mM MgCl2 is also provided. Store at -20°C. Guaranteed stable for 6 months when properly stored.

Guaranteed minimum shelf life is 60 days (exact expiry date printed on product and CofA).

Documents

Manuals & protocols

Recommended products