This product will be discontinued by June 2017. View alternative products >

Platinum® Quantitative RT-PCR ThermoScript™ One-Step System

Catalog number: 11731023

Invitrogen™  Related applications: Real Time PCR (qPCR) | Real Time PCR-Based Gene Expression Profiling

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

NOTIFICATION: Platinum™ Quantitative RT-PCR ThermoScript™ One-Step System will be discontinued on June 30th, 2017. Try TaqMan® Fast Virus 1-Step Master Mix for superior performance.
We encourage you to plan your needs for the next year and request your last orders by the discontinuation date.

Platinum Quantitative RT-PCR ThermoScript One-Step System is for real-time quantitation of RNA molecules from total or poly(A)+ RNA in a single step.

This kit provides:
• Platinum™ Taq DNA Polymerase
• ThermoScript RT polymerase
• A vial of 50 mM MgSO4

For Research Use Only. Not for use in diagnostic procedures.

Specifications

PCR Method: 1-Step qRT-PCR
Detection Method: Primer-Probe Detection
Reaction Speed: Standard
Sample Type (General): RNA
Product Line: Platinum®, ThermoScript™
Product Size: 500 reactions
Shipping Condition: Dry Ice

Contents & storage

Platinum® Quantitative RT-PCR ThermoScript™ One-Step System contains ThermoScript™ RT/Platinum® Taq mix, and 2X ThermoScript™ reaction mix (a buffer containing 0.4 mM of each dNTP, 6 mM MgSO4 ). A vial of 50 mM MgSO4 is also provided. Store at -20°C. Guaranteed stable for 6 months when properly stored.

Documents

Manuals & protocols