Invitrogen™

SuperScript™ III Platinum™ One-Step qRT-PCR Kit

Catalog number:  11732020

 Related applications: Real Time PCR (qPCR) | Real Time PCR-Based Gene Expression Profiling

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The SuperScript® III Platinum® One-Step qRT-PCR Kits combine the most powerful reverse transcriptase and DNA polymerase technologies to deliver precise and accurate analysis of gene expression in a convenient one-step format. The SuperScript® III Platinum® One-Step qRT-PCR Kit provides sensitive, specific detection with fluorogenic primers and probes, while the SuperScript® III Platinum® SYBR® Green One-Step qRT-PCR Kit offers the ease and convenience of SYBR® Green I detection

SuperScript® III Platinum® One-Step qRT-PCR Kits provide:

• SuperScript® III RT for higher temperature cDNA synthesis for greater success with difficult RNA secondary structure
• Platinum® Taq DNA Polymerase with hot-start technology for improved specificity
• LUX™ Fluorogenic Primers (Figure 1) or dual-labeled fluorogenic probes (Figure 2) for superior detection performance
• SYBR® Green I dye for easy and convenient detection (Figure 3)
• Validated performance on multiple real-time instrument platforms, including rotor-based systems
For Research Use Only. Not for use in diagnostic procedures.

Specifications

PCR Method: 1-Step qRT-PCR
Detection Method: Primer-Probe Detection
Reaction Speed: Standard
For Use With (Equipment): 7500 System, BioRad MyiQ, BioRad iCycler iQ, BioRad iQ5, Cepheid SmartCycler, MJ Chromo4, MJ Opticon, Stratagene Mx3000P, Stratagene Mx3005P, Stratagene Mx4000
Sample Type (General): RNA
Product Line: Platinum®, SuperScript™
Product Size: 100 reactions
Shipping Condition: Dry Ice

Contents & storage

• 100 µl SuperScript® III/Platinum® Taq Mix
• 2 × 1.25 ml 2X Reaction Mix
• 1 ml Magnesium Sulfate (50 mM)
• 100 µl ROX Reference Dye (25 µM)

Store components at -20°C. ROX Reference Dye must be stored in the dark.

Documents

Manuals & protocols

Recommended products