Platinum® SYBR® Green qPCR SuperMix-UDG

Catalog number: 11733046

Invitrogen™  Related applications: Chromatin Biology | Real Time PCR (qPCR) | Real Time PCR-Based Gene Expression Profiling

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

**********************
Alternative Product: Try PowerUp SYBR Green Master Mix, our newest, high-performance, SYBR dye-based master mix for superior performance at a very competitive price. With PowerUp SYBR Green Master Mix, we’ve taken the best of Platinum SYBR Green qPCR SuperMix-UDG and added additional capabilities for your gene expression analysis.

**********************
Platinum® SYBR® Green qPCR SuperMix-UDG provides a sensitive and convenient system for real-time PCR detection with SYBR® Green I dye. The SuperMix format combines amplification and detection reagents into a ready-to-go mix to make set-up easy and ensure reproducible results. Platinum® SYBR® Green qPCR SuperMix-UDG offers:

–Platinum® Taq DNA Polymerase for sensitive amplification from as few as 10 copies of target (Figure 1)

–100% dUTP and UDG enzyme for superior control of carryover contamination, saving time by reducing the number of failed experiments
–Separate tubes of ROX reference dye and BSA for easy optimization on 96-well plates and glass capillary tube instruments (Figure 2)
–SYBR® Green I dye for simple and easy detection of real-time product
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Detection Method: SYBR
For Use With (Equipment): 7000 System, 7300 System, 7500 System, 7700 System, 7900HT System, BioRad iCycler iQ, Cepheid SmartCycler, GeneAmp® 5700, StepOne™, StepOnePlus™, ViiA™ 7 System
Reaction Speed: Standard
PCR Method: qPCR
Sample Type (General): DNA
Product Line: Platinum®, SYBR®
Product Size: 500 reactions
Sample Type (Specific): DNA (Genomic), cDNA
Shipping Condition: Dry Ice

Contents & storage

• Platinum® SYBR® Green qPCR SuperMix-UDG (12.5 mL)
• 50 mM Magnesium Chloride (2 × 1 mL)
• ROX Reference Dye (500 µL)
• 20X BSA (1.3 mL)

Sufficient reagents are provided for 500 reactions of 50 µL each. Components may be stored at either -20°C or +4°C.

Documents

Manuals & protocols

Recommended products