SYBR GreenER™ qPCR SuperMix for ABI PRISM® Instrument

Catalog number: 11760500

Invitrogen™  Related applications: Real Time PCR (qPCR) | Real Time PCR-Based Gene Expression Profiling

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

**********************
Alternative Product: Try PowerUp SYBR Green Master Mix, our newest, high-performance, SYBR dye-based master mix for superior performance at a very competitive price. With PowerUp SYBR Green Master Mix, we’ve taken the best of SYBR GreenER qPCR SuperMix and added additional capabilities for your gene expression analysis.

**********************
SYBR® GreenER™ qPCR SuperMixes for ABI PRISM® incorporate the latest high-performance technology to provide the most reliable gene expression data from your ABI 7300, 7000, 7700 or 7900HT instrument. The SYBR® GreenER™ system is specially formulated to offer the best sensitivity, specificity, and reproducibility with:
• A novel dsDNA-binding dye - exhibits a brighter signal for increased sensitivity and less PCR inhibition than original SYBR® Green I, while using the same instrument filters and settings
• An optimized buffer system - improves sensitivity and provides excellent long-term stability
• Uracil DNA glycosylase - reduces carryover contamination in qPCR

Novel Formulation Provides More Reliable Data
The SYBR® GreenER™ qPCR SuperMix kits include a new dsDNA-binding dye that produces a brighter signal and significantly less PCR inhibition, for improved qPCR performance over a broad dynamic range.

Consistent Specificity and Low-Copy Detection
The SYBR® GreenER™ qPCR reagent system minimizes the formation of nonspecific products, including primer-dimers, for greater accuracy across a wide range of targets. The reliability of the SYBR® GreenER™ qPCR reagent system also translates to greater sensitivity. With SYBR® GreenER™ qPCR SuperMix, you can consistently detect fewer than 10 copies of a target in genomic DNA.

For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Detection Method: SYBR
For Use With (Equipment): 7000 System, 7300 System, 7700 System, 7900HT Fast System, 7900HT System
Reaction Speed: Standard
PCR Method: qPCR
Sample Type (General): DNA
Product Line: Platinum®, SYBR® GreenER™
Product Size: 500 reactions
Sample Type (Specific): DNA (Genomic), cDNA
Shipping Condition: Dry Ice

Contents & storage

Contains SYBR® GreenER™ qPCR SuperMix for ABI PRISM® (12.5 mL). SYBR® GreenER™ qPCR SuperMix for ABI PRISM® is a 2X SuperMix with premixed ROX Reference dye. Sufficient reagents are provided for 500 reactions based on a 50 µL reaction size.

Store at +4°C upon receipt.

Documents

Manuals & protocols

Recommended products