This product will be discontinued by December 2016. View alternative products >

EXPRESS One-Step SYBR® GreenER™ Kit, universal

Catalog number: 11780200

Invitrogen™  Related applications: Real Time PCR (qPCR) | Real Time PCR-Based Gene Expression Profiling

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

NOTIFICATION: EXPRESS One-Step SYBR™ GreenER™ kits will be discontinued on June 30th, 2017. Try Power SYBR™ Green RNA-to-CT™ 1-Step Kit, our highest-performance, SYBR dye-based master mix for superior performance at a very competitive price.
We encourage you to plan your needs for the next year and request your last orders by the discontinuation date.

EXPRESS One-Step SYBR GreenER kits are compatible with fast-cycling instruments for your SYBR-based qPCR experiments. This kit comes with a separate tube of ROX reference dye. EXPRESS One-Step SYBR GreenER SuperMix kits with premixed ROX reference dye are also available.

This kit provides:
• Fast-activating, antibody-mediated Platinum™ Taq DNA Polymerase
• The One-Step SuperScript™ III Reverse Transcriptase for one-step qRT-PCR
• SYBR GreenER dye
• UDG carryover protection—uracil-DNA-glycosylase (UDG)/dUTP to reduce carryover contamination

For Research Use Only. Not for use in diagnostic procedures.

Specifications

PCR Method: 1-Step qRT-PCR
Detection Method: SYBR
Reaction Speed: Fast, Standard
For Use With (Equipment): 7500 Fast System, 7500 System, BioRad CFX96™, BioRad MyiQ, BioRad iCycler iQ, Cepheid SmartCycler, Corbett RotorGene, MJ Chromo4, MJ Opticon, StepOnePlus™, Fast Mode, StepOnePlus™, Standard Mode, Stratagene Mx3000P, Stratagene Mx3005P, Stratagene Mx4000
Sample Type (General): RNA
Product Line: SYBR® GreenER™
Product Size: 500 reactions

Contents & storage



EXPRESS One-Step SYBR® GreenER™ SuperMix Kits are composed of 2 modules: EXPRESS SYBR® GreenER™ RT module (containing SuperScript® III Reverse Transcriptase and RNAseOUT™) and EXPRESS SYBR® GreenER™ qPCR SuperMix with ROX in a separate tube.

Store at -20°C upon receipt. Once thawed, the EXPRESS SYBR® GreenER™ qPCR SuperMix can be stored for up to 1 month at +4·C. Product is guaranteed to be stable for 6 months when properly stored.

Documents

Manuals & protocols

Recommended products