This product will be discontinued by June 2017. View alternative products >

EXPRESS SYBR™ GreenER™ qPCR Supermix, universal

Catalog number: 1178401K

Invitrogen™  Related applications: Real Time PCR (qPCR) | Real Time PCR-Based Gene Expression Profiling

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

NOTIFICATION: EXPRESS SYBR™ GreenER™ qPCR SuperMix will be discontinued on June 30th, 2017. Try PowerUp™ SYBR™ Green Master Mix, our newest, high-performance, SYBR dye-based master mix for superior performance at a very competitive price.
We encourage you to plan your needs for the next year and request your last orders by the discontinuation date.

EXPRESS SYBR GreenER qPCR SuperMix kits are compatible with fast-cycling instruments for your SYBR-based qPCR experiments. EXPRESS SYBR GreenER qPCR SuperMix Universal comes with separate tubes of ROX reference dye. EXPRESS SYBR GreenER qPCR SuperMix with Premixed ROX is also available.

This kit provides:
• Fast-activating, antibody-mediated Platinum™ Taq DNA Polymerase
• SYBR GreenER dye
• UDG carryover protection—uracil-DNA-glycosylase (UDG)/dUTP to reduce carryover contamination

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Detection Method: SYBR
For Use With (Equipment): 7500 Fast System, 7500 System, BioRad CFX96™, BioRad MyiQ, BioRad iCycler iQ, Cepheid SmartCycler, Corbett RotorGene, MJ Chromo4, MJ Opticon, StepOnePlus™, Fast Mode, StepOnePlus™, Standard Mode, Stratagene Mx3000P, Stratagene Mx3005P, Stratagene Mx4000
Reaction Speed: Fast, Standard
PCR Method: qPCR
Sample Type (General): DNA
Product Line: SYBR® GreenER™
Product Size: 1,000 reactions
Sample Type (Specific): DNA (Genomic), cDNA

Contents & storage

Contains 5 × 5 ml EXPRESS SYBR® GreenER™ qPCR SuperMix Universal (2X) and 5 × 500 µl ROX Reference Dye. Sufficient reagents are provided for 2500 reactions of 20 µL each.

Store all components at -20°C for long-term storage. EXPRESS qPCR SuperMixes may be stored at 4–8°C for up to one month.

Documents

Manuals & protocols

Recommended products