EXPRESS qPCR Supermix, universal

Catalog number:  11785200

Invitrogen™  Related applications: Real Time PCR (qPCR) | Real Time PCR-Based Gene Expression Profiling

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Alternative Product: Try TaqMan Fast Advanced Master Mix, our highest-performance, probe-based master mix. With TaqMan Fast Advanced Master Mix, we’ve taken the best of EXPRESS qPCR SuperMix and added additional capabilities for your gene expression analysis.

EXPRESS qPCR SuperMix kits are compatible with fast-cycling instruments for your probe-based qPCR assays. This kit comes with a separate tube of ROX reference dye. EXPRESS qPCR SuperMix kits with premixed ROX reference dye are also available.

This kit provides:
• Fast-activating, antibody-mediated Platinum™ Taq DNA Polymerase
• UDG carryover protection—uracil-DNA-glycosylase (UDG)/dUTP to reduce carryover contamination

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Detection Method: Primer-Probe Detection
For Use With (Equipment): 7500 Fast System, 7500 System, BioRad CFX96™, BioRad MyiQ, BioRad iCycler iQ, Cepheid SmartCycler, Corbett RotorGene, Eppendorf® MasterCycler® RealPlex, MJ Chromo4, MJ Opticon, StepOnePlus™, Fast Mode, StepOnePlus™, Standard Mode, Stratagene Mx3000P, Stratagene Mx3005P, Stratagene Mx4000
Reaction Speed: Fast, Standard
PCR Method: qPCR
Sample Type (General): DNA
Product Size: 500 reactions
Sample Type (Specific): DNA (Genomic), cDNA

Contents & storage



EXPRESS qPCR SuperMix kits are provided as a 2X qPCR SuperMix with ROX premixed or in a separate tube.

Store at -20°C upon receipt. Once thawed, the EXPRESS qPCR SuperMix can be stored for up to 1 month at +4·C. Product is guaranteedstable for 6 months when properly stored.

Documents

Manuals & protocols

Recommended products