New! Save technical documents to your personal library. Open a PDF, then look for the "Save to Library" icon.

EXPRESS One-Step Superscript™ qRT-PCR Kit, with premixed ROX

Catalog number:  11791200

Invitrogen™  Related applications: Real Time PCR (qPCR) | Real Time PCR-Based Gene Expression Profiling

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top


Next-generation, real-time qRT-PCR reagents designed to complement fast-cycling instruments
Get the turnaround time you want with the data quality you need

EXPRESS One-Step SuperScript® qRT-PCR SuperMix Kits incorporate proven Invitrogen enzymes now specifically tailored to deliver superior results on high-throughput, fast-cycling instruments for your probe-based qPCR experiments. These kits provide:

• Fast-activating, antibody-mediated Platinum® Taq DNA Polymerase - complete, rapid activation provides the best sensitivity and reproducibility (Figure 1)
• The One-Step SuperScript® III Reverse Transcriptase module has a unique formulation to ensure better yield and less PCR inhibition, specifically for one-step qRT-PCR
• Flexible formats with ROX premixed or separate - designed to work with default instrument protocols on a wide range of high-throughput fast instruments (e.g., AB 7500 Fast, AB 7900 HT Fast, AB StepOne, Roche LightCycler® 480, Corbett Rotor-Gene™) (Figure 2)
• The only commercially available one-step qRT-PCR kits containing heat-labile UDG - significantly reduced carryover contamination risk with excellent activity from room temperature to approximately 40·C. Due to the thermostability of SuperScript® III, the cDNA synthesis step is run at 50·C, at which temperature the heat-labile UDG is totally and irreversibly inactivated (Figure 3)

Easy to test and switch

EXPRESS kits work with manufacturer’s default protocols on fast instruments and come in a convenient and stable SuperMix format for ultimate convenience. The kits are offered with either premixed ROX or a separate tube of ROX to fit your instrument needs.
For Research Use Only. Not for use in diagnostic procedures.


PCR Method: 1-Step qRT-PCR
Detection Method: Primer-Probe Detection
Reaction Speed: Fast, Standard
For Use With (Equipment): 7000 System, 7300 System, 7700 System, 7900HT Fast System, 7900HT System, Eppendorf® MasterCycler® RealPlex, StepOne™, Fast Mode, StepOne™, Standard Mode
Sample Type (General): RNA
Product Line: SuperScript™
Product Size: 500 reactions

Contents & storage

EXPRESS One-Step SuperScript® qRT-PCR SuperMixes are composed of 2 modules: an EXPRESS RT module (containing SuperScript® III Reverse Transcriptase and RNAseOUT™) and an EXPRESS qPCR SuperMix module with ROX premixed or in a separate tube.

Store at -20°C upon receipt. Once thawed, the EXPRESS qPCR SuperMix can be stored for up to 1 month at +4·C. Product is guaranteed stable for 6 months when properly stored.


Manuals & protocols