Kanamycin Sulfate

Catalog number: 11815024

Gibco™  Related applications: Transformation

Error loading your content!

  Catalog number
Select a plan
Unit size
List price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Kanamycin Sulfate is a water-soluble antibiotic originally purified from the bacterium Streptomyces kanamyceticus. Kanamycin acts by binding to the 30S subunit of the bacterial ribosome and inhibiting protein synthesis in susceptible bacteria. Gibco® Kanamycin is effective against a wide variety of gram-negative and some gram-positive bacteria, and is used for the prevention of bacterial contamination of cell cultures. We offer a variety of antibiotics and antimycotics for cell culture applications.

Product Use
For Research Use Only: Not intended for animal or human diagnostic or therapeutic use.

Dual-Site cGMP Manufacturing
For supply chain continuity, we manufacture Gibco® Kanamycin at two separate facilities located in Grand Island, NY and Scotland, UK. Both sites are compliant with cGMP manufacturing requirements, are certified to ISO 13485, and are registered with the FDA as medical device manufacturers.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Validated Application: Bacterial Selection
Agent: Kanamycin
Form: Powder
Reagent Type: Antibiotic (Selective)
Product Size: 5 g
Shipping Condition: Room Temperature

Contents & storage

Store at room temperature.

Documents

Manuals & protocols

Recommended products