New! Save technical documents to your personal library. Open a PDF, then look for the "Save to Library" icon.
Invitrogen™

Gateway™ Vector Conversion System with One Shot™ ccdB Survival Cells

Catalog number:  11828029

 Related applications: Gateway Cloning

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The Gateway® Vector Conversion System is designed to convert any vector into a Gateway® destination vector using restriction endonucleases and ligase. A Gateway® cassette containing attR recombination sites flanking a ccdB gene (1) and a chloramphenicol-resistance gene are blunt-end cloned into the multiple cloning site of any vector. ccdB Survival 2 competent cells allow propagation of the Gateway® vectors containing the ccdB gene. The Gateway® Vector Conversion System offers:
• Conversion of any expression vector into a Gateway® destination vector
• Three cassettes to construct a destination vector in the correct reading frame. Each cassette has a unique restriction site to easily distinguish the cassettes and screen for the correct orientation.
For Research Use Only. Not for use in diagnostic procedures.

Figures

Specifications

Antibiotic Resistance (Bacterial): Chloramphenicol (CmR)
Bacterial or Yeast Strain: ccdB Survival™ 2
Cloning Method: Gateway®
Product Line: Gateway®, One Shot®
Product Size: 1 kit

Contents & storage

The Gateway® Vector Conversion System contains Gateway® Reading Frame Cassettes A, B, and C.1, pENTR™-gus positive control, and One Shot® ccdB Survival Cells. Store DNA at -20°C and competent cells at -80°C.

Documents

Manuals & protocols

Recommended products