SuperScript® One-Step RT-PCR System for Long Templates

Catalog number: 11922010

Invitrogen™  Related applications: Reverse Transcription

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The SuperScript® One-Step RT-PCR for Long Templates is designed for the convenient, sensitive, and reproducible detection and analysis of RNA molecules by one-step RT-PCR. Both cDNA synthesis and PCR are performed in a single tube using gene-specific primers and specific target RNAs from either total RNA or mRNA. Due to the high fidelity of the DNA polymerase blend in the enzyme mix, this kit can also be used for one-step cloning of genes.

Using the SuperScript® One-Step System
The system consists of two major components: SuperScript® II RT/Platinum® Taq Hi Fi Mix and 2X Reaction Mix. The enzyme mix is a mixture of SuperScript® II Reverse Transcriptase, which provides reduced RNase H activity and increased thermal stability, and Platinum® Taq DNA Polymerase High Fidelity, which is a blend of recombinant Taq DNA polymerase and the proofreading enzyme Pyrococcus species GB-D thermostable polymerase. The Taq DNA polymerase in the blend is complexed with a proprietary antibody that inhibits enzyme activity at ambient temperatures. Activity is regained after the denaturation step in PCR cycling at 94°C, providing an automatic "hot start" for Taq DNA polymerase in PCR. Hot starts in PCR provide increased sensitivity, specificity, and yield, while allowing assembly of reactions at ambient temperatures. The 2X Reaction Mix consists of a proprietary buffer system optimized for reverse transcription and PCR amplification, Mg2+ optimized for universal use, deoxyribonucleotide triphosphates, and stabilizers. This convenient 2X format allows further addition of template and primer at any desired concentration. Sufficient reagents are provided for 100 amplification reactions of 50 µL each. This enzyme mixture can amplify RNA targets up to 9.0 kb.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

PCR Method: 1-Step RT-PCR
Reverse Transcriptase: SuperScript™ II
Polymerase: Platinum® Taq DNA Polymerase High Fidelity
Hot Start: Built-In Hot Start
Sample Type (General): RNA
Detection Method: Gel Electrophoresis
Final Product Size(s): 9 kb or less
Primer-Probe Compatibility: Unlabeled Oligos
Product Line: Platinum®, SuperScript®
Product Size: 25 reactions
Shipping Condition: Wet Ice

Contents & storage

SuperScript® One-Step RT-PCR System components are listed in Table 1. Store at -20°C. Guaranteed stable for 6 months when properly stored.

Documents

Manuals & protocols

Recommended products