Hybridoma-SFM

Catalog number: 12045084

Gibco™  Related applications: Antibody & Cell Culture Production | Mammalian Cell Culture

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Hybridoma-SFM is a serum-free, very low-protein medium specifically developed to support the growth of hybridomas for antibody production. Hybridoma-SFM features:

• Ability to support suspension and stationary hybridomas and myelomas
• Complete, ready-to-use formulation
• Serum-free, very low-protein formulation

Ability to support suspension and stationary hybridomas and myelomas
Suitable for the culture of recombinant myeloma lines as well as traditional hybridomas, Hybridoma-SFM provides high cell densities and IgG production. Cholesterol-dependent cultures such as NS0 require the addition of 250X Cholesterol Lipid Concentrate.

Complete, ready-to-use formulation
Hybridoma-SFM is a complete medium that is ready to use. No supplementation is required.

Serum-free, very low-protein formulation
Hybridoma-SFM is a serum-free, very low-protein formulation, allowing for easier purification of your protein of interest. Hybridoma-SFM contains <20 µg/mL total protein.

Product use
Customers using Gibco® Hybridoma-SFM in a manufacturing process, who have a submission with the FDA, may request a letter of authorization from us to reference our Type II Drug Master File (DMF).

cGMP manufacturing and quality system
Hybridoma-SFM is manufactured at a cGMP-compliant facility located in Grand Island, New York. The facility is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Form: Liquid
Cell Type: Hybridomas
Glutamine: L-Glutamine
Serum Level: Serum-Free
Product Size: 500 mL
Classification: Serum-Free, Low-Protein
Phenol Red Indicator: Phenol Red
Shipping Condition: Room Temperature

Contents & storage

Storage conditions: 2-8°C. Protect from light
Shipping conditions: Ambient
Shelf life: 12 months from date of manufacture

Documents

Manuals & protocols

Recommended products