Gibco™

CHO-S-SFM II, with hypoxanthine and thymidine This product, and/or its packaging, incorporates one or more environmental/green features. Click on this icon to learn more.

Catalog number:  12052098

 Related applications: Antibody & Cell Culture Production | Mammalian Cell Culture

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

CHO-S-SFM II, is a complete, serum-free, low-protein (less than 100 µ/ml) medium optimized for the growth and maintenance of Chinese hamster ovary (CHO) cells in suspension culture for production of recombinant proteins. It is supplemented with hypoxanthine and thymidine (the non-supplemented equivalent is also available). This ready-to-use medium is suitable for the suspension culturing of CHO and other cells in batch, continuous, and perfusion culture systems. A CHO-S-SFM II medium Drug Master File (DMF) has been submitted to the FDA. Permission to cross-reference the Master File may be obtained by contacting Technical Services or your local Sales Representative.

This medium contains components of animal origin. If growth factors or hydrolysates, found in competitor media, are to be avoided, consider CD CHO, CD OptiCHO™ or CD FortiCHO™ media.

When to use CHO-S-SFM II Medium
• Your cells have been successfully cultured in CHO-S-SFM II medium in the past and your process is nearing a regulatory filing (DMF available)

When to Consider Another Gibco® Medium

Consider CD FortiCHO™ medium when:
• The cell line you are using is a transfected CHO K1, GS CHO or CHO-S™ cell line
• Maximum batch culture cell densities and protein titers are needed
• You have time to adapt cells into a new medium in an effort to maximize titers
• Cell health at harvest is a priority for downstream processing

Consider CD OptiCHO™ medium when:
• You are using a transfected CHO cell line other than CHO K1, GS CHO or CHO-S™ cell lines
• Optimization of feeding strategies will be part of your base medium selection testing in an effort to maximize productivity
• Your CHO cell line is “finicky," i.e. hard to grow or adapt

Consider CD CHO medium when:
• Your GS CHO cells are recommended to be grown in CD CHO medium
• Your cells have been successfully cultured in CD CHO medium in the past and your process is nearing a regulatory filing (DMF available)

Consider CD DG44 medium when:
• You use parental DG44 or DXB11 CHO cells (dhfr-)

Design Your Own Media Specific to Your Needs
Whether it’s a bench-level experiment or large scale biomanufacturing process, we offer specialized formats, packaging, and quality levels specific to your needs. With Gibco® cGMP Media Custom Services you can put the manufacturing power and years of technical experience of our R&D and Bioproduction Application Specialists to work for you.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Cell Line: CHO
Classification: Low-Protein, Serum-Free
Concentrated: 1 X
Culture Type: Suspension Cell Culture
Endotoxin Level: Low
Form: Liquid
Format: Bottle(s)
Glutamine: L-Glutamine
Included Antibiotics: No antibiotics
Phenol Red Indicator: No Phenol Red
Product Line: Gibco®
Product Size: 1,000 mL
Serum Level: Serum-Free
Sodium Bicarbonate Buffer: Sodium Bicarbonate
Sodium Pyruvate Additive: Sodium Pyruvate
Species: Hamster
Supplements: Hypoxanthine, Thymidine
Volume (Metric): 1 L
Green Features: Sustainable packaging
Shipping Condition: Room Temperature

Contents & storage

Store in refrigerator (2–8°C) and protect from light.

Documents

Manuals & protocols

Recommended products