This product is manufactured in our Grand Island, US facility and is intended for sale in North America, Latin America, and the Asia/Pacific region—higher freight charges will apply to orders shipped elsewhere. An equivalent product, manufactured in Paisley, Scotland, is recommended for our customers in Europe, Africa, and the Middle East. Click to view.

IMDM, powder

Catalog number:  12200069

Gibco™  Related applications: Mammalian Cell Culture

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top


Iscove's Modified Dulbecco's Medium (IMDM) is well suited for rapidly proliferating, high-density cell cultures, including Jurkat, COS-7, and macrophage cells. We offer a variety of Gibco® IMDM modifications for a range of cell culture applications. Find the right formulation using the media selector tool.

This IMDM is modified as follows:

With Without
• L-glutamine • α-thioglycerol
• Phenol Red • 2-mercaptoethanol

The complete formulation is available.

IMDM, a modification of Dulbecco's Modified Eagle Medium, includes selenium as well as additional amino acids and vitamins. In addition, this unique medium lacks iron, with potassium nitrate replacing ferric nitrate.

Product Intended Use
For in vitro diagnostic use. CAUTION: Not for human or animal therapeutic use. Uses other than the intended use may be a violation of local law.

cGMP Manufacturing and Quality System
Gibco® IMDM is manufactured at a cGMP compliant facility, located in Grand Island, New York. The facility is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards. For supply chain continuity, we offer an identical Gibco® IMDM product made in our Scotland facility (42200-030). This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard.

IMDM contains no proteins, lipids, or growth factors. Therefore, IMDM requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). IMDM uses a sodium bicarbonate buffer system (3.024 g/L), and therefore requires a 5-10% CO2 environment to maintain physiological pH.

Powder forms of Gibco® cell culture medium require sodium bicarbonate supplementation, pH adjustment, and filtration at the time of preparation (see protocol for details).


Glutamine: L-Glutamine
Phenol Red Indicator: Phenol Red
Form: Powder
Sodium Pyruvate Additive: Sodium Pyruvate
Glucose: High Glucose
Product Line: Gibco®
Product Size: 10 L
Shipping Condition: Room Temperature

Contents & storage

Storage conditions: 2-8° C
Shipping conditions: Ambient
Shelf life: 36 months from date of manufacture

Media formulations


Manuals & protocols

Recommended products