ThermoScript™ Reverse Transcriptase

Catalog number: 12236014

Invitrogen™  Related applications: Reverse Transcription

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

ThermoScript™ Reverse Transcriptase (RT) is an avian RT with reduced RNase H activity designed for RT-PCR (1). It is engineered to have high thermal stability (up to 70°C) (Figure 1), copy RNA with secondary structure, and improve priming specificity (2).

Yield—Reduced RNase H activity results in greater first-strand cDNA yields
Size of cDNA—Generation of RT-PCR products up to 12 kb
Thermostability—ThermoScript™ Reverse Transcriptase is active up to 70°C
Flexibility—Uses either total or poly(A)+ RNA

Unit Definition: One unit incorporates 1 nmol of dTTP into acid-precipitable material in 10 min at 37°C using poly(A)-oligo(dT)25 as template-primer.

For Research Use Only. Not intended for any animal or human therapeutic or diagnostic use.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Optimal Reaction Temperature: 65° C
Final Product Size(s): 12.3 kb or less
Reverse Transcriptase: ThermoScript™
Ribonuclease H Activity: Reduced
Form: Frozen
Format: Tube(s)
Quantity: 375 units
Template: ssRNA
Sensitivity: Medium
Product Size: 375 units
Concentration: 15 U&⁄µl
Enzyme Function: RNA dependent DNA Polymerase
Shipping Condition: Dry Ice
Number of Reactions: 25 Reactions

Contents & storage

ThermoScript™ RT is supplied with a vial of 5X cDNA synthesis buffer [250 mM Tris acetate (pH 8.4), 375 mM potassium acetate, 40 mM magnesium acetate, and stabilizers], and a vial of 100 mM DTT. Store at -70°C. Guaranteed stable for 6 months when properly stored.

Documents

Manuals & protocols

Recommended products