Invitrogen™

SequenceRx Enhancer Solution A

Catalog number:  12238010

 Related applications: Sanger Sequencing

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The SequenceRx Enhancer System is designed to improve the length and quality of sequencing data from difficult sequencing templates. It is a panel of seven co-solvents which are added individually to automated fluorescent DNA sequencing reactions. This system is not designed for use on long stretches of poly A.

The SequenceRx Enhancer System:

• Is useful on templates containing regions with secondary structure such as high GC-content, small nucleotide repeats, hairpins, direct repeats, and inverted repeats
• Requires no change in protocols-the SequenceRx Solution replaces part of the water added to the reaction mixture
• Is compatible with all sequencing chemistries and platforms
• Uses one reaction to determine the sequence for all four nucleotides (100 reactions equal 100 templates)
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Compatible Products: BigDye® terminator products, Rhodaminedye terminator products, dRhodamine dye terminator products
For Use With (Equipment): Sequencing Instruments (Capillary Electrophoresis-Based)
Product Size: 200 reactions
Shipping Condition: Approved for shipment on Wet or Dry Ice

Contents & storage

The SequenceRx Enhancer System includes the seven reagents listed in Table 1. Solutions are also available separately. Store at -20°C. Guaranteed stable for 6 months when properly stored.

Documents

Manuals & protocols

Recommended products