Gibco™

MarrowMAX™ Bone Marrow Medium This product, and/or its packaging, incorporates one or more environmental/green features. Click on this icon to learn more.

Catalog number:  12260014

 Related applications: Clinical & Translational Research | Mammalian Cell Culture | Stem Cell Culture

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Gibco® MarrowMAX™ Bone Marrow Medium is a fully-supplemented medium developed for the short term culture of bone marrow and other hematopoietic cells for cytogenetic studies and in vitro diagnostic procedures.

Gibco® MarrowMAX™ Bone Marrow Medium features:

• Superior performance
• Ready-to-use format
• Quality and performance testing
• Unique, optimized formulation

Superior performance
Gibco® MarrowMAX™ Bone Marrow Medium outperforms other commercially available media containing Giant Cell Tumor (GCT) conditioned media, providing consistent performance with a higher mitotic index and superior chromosomal morphology (learn more about the superior performance of MarrowMAX™ medium here).

Ready-to-use format
Gibco® MarrowMAX™ Bone Marrow Medium is a frozen, 1X medium, ready to use upon thawing. No supplementation is required. Thawed medium can be stored at 2–8°C for up to 60 days.

Quality and performance testing
Every lot of Gibco® MarrowMAX™ Bone Marrow Medium is performance tested by a certified US reference cytogenetics laboratory to ensure consistently superior performance. Bone marrow cells from the aspirate of a normal adult donor are cultured for 24 hours in Gibco® MarrowMAX™ Bone Marrow Medium before measuring the mitotic index and chromosome banding resolution. In addition, each lot is tested for sterility, pH, osmolality, mycoplasma and endotoxin.

Unique, optimized formulation
Gibco® MarrowMAX™ Bone Marrow Medium is an optimized RPMI 1640 medium, supplemented with Fetal Bovine Serum (FBS), gentamicin, and L-glutamine. It also contains a novel human stromal cell conditioned medium that consists of a unique blend of hematopoietic growth factors. Gibco® MarrowMAX™ Bone Marrow Medium does not contain GCT conditioned medium.

Product Use
For in vitro diagnostic use.

cGMP Manufacturing and Quality System
Gibco® MarrowMAX™ Bone Marrow Medium is manufactured at a cGMP compliant facility, located in Grand Island, New York. The facility is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards.

Specifications

Cell Type: Bone Marrow, Hematopoietic Cells
Form: Liquid
Glutamine: L-Glutamine
Product Line: MarrowMAX™
Product Size: 100 mL
Serum Level: Standard Serum
Species: Human
Supplements: Conditioned Medium (from Stromal Cells), FBS
Green Features: Sustainable packaging
Shipping Condition: Dry Ice

Contents & storage

Storage conditions: -5°C to -20°C. Protect from light.
Shipping conditions: Frozen
Shelf life: 18 months from date of manufacture. Thawed medium can be stored at 2°C to 8°C for up to 60 days.

Documents

Manuals & protocols

Recommended products