New! Save technical documents to your personal library. Open a PDF, then look for the "Save to Library" icon.
Invitrogen™

Gateway™ pcDNA™-DEST40 Vector

Catalog number:  12274015

 Related applications: Gateway Cloning | Protein Expression

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The pcDNA™ vectors are designed for high-level, constitutive expression in a variety of mammalian cell lines. The Gateway® pcDNA™-DEST40 vector offers the following key features:

•C-terminal V5-6x His tag for easy detection and rapid purification
•Cytomegalovirus (CMV) promoter for high-level expression
attR sites for Gateway® cloning, enabling recombination with attL-flanked fragments
•Neomycin resistance gene for stable selection
•Ampicillin resistance gene and pUC origin for selection and maintenance in E. coli

Gateway® Cloning
To fit all of your expression needs, Invitrogen offers state-of-the-art Gateway® destination vectors for expression in E. coli, insect, yeast, or mammalian cells, as well as for production of native protein or N- or C-terminal fusion proteins. All Gateway® destination vectors have attR sites for recombination with any attL-flanked fragment, regardless of whether it is an entry clone or an Ultimate™ ORF Clone. The following table lists a variety of available destination vectors.

Additional materials required, available separately: Gateway® entry clone, appropriate Gateway® LR Clonase® enzyme mix, and reaction buffer.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Promoter: CMV
Key Function: Constitutive Expression
Protein Tag or Fusion: His Tag (6x), V5 Epitope Tag
Delivery Method: Transfection
Cloning Method: Gateway®
Selection Agent (Eukaryotic): Geneticin® (G-418)
Vector Type: pDEST
Constitutive or Inducible System: Constitutive
Product Line: Gateway®, pcDNA™
Product Size: 6 µg

Contents & storage

All destination vectors are provided lyophilized and supercoiled.

Documents

Manuals & protocols

Recommended products