Invitrogen™

pBAD-DEST49 Gateway™ Destination Vector

Catalog number:  12283016

 Related applications: Gateway Cloning | Protein Expression

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The pBAD-DEST49 Gateway® destination vector is designed for rapid cloning with a Gateway® entry clone using lambda phage site-specific recombination and subsequent high-level, tightly regulated expression in E. coli. The pBAD-DEST49 vector features:

• The araBAD promoter for tightly regulated expression in E. coli
• HP-thioredoxin fusion for improved protein yield and solubility
• Enterokinase cleavage site for efficient cleavage of the N-terminal fusion with EKMax™
• C-terminal V5 and 6xHis tags for efficient detection and purification of fusion proteins
• R sites for efficient recombination with any attL-flanked Gateway® entry vector
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Vector Type: pBAD, pDEST
Cloning Method: Gateway®
Promoter: araBAD
Protein Tag or Fusion: His Tag (6x), Thioredoxin, V5 Epitope Tag
Antibiotic Resistance (Bacterial): Ampicillin (AmpR)
Cleavage: EK (Enterokinase) Recognition Site
Constitutive or Inducible System: Inducible
Inducing Agent: Arabinose
Product Line: Gateway®
Product Size: 6 µg
Selection Agent (Eukaryotic): None

Contents & storage

The pBAD-DEST49 Gateway® destination vector includes 6 µg of vector. Store at -20°C. The vector is guaranteed stable for 6 months when properly stored.

Documents

Manuals & protocols