Gibco™

293fectin™ Transfection Reagent

Catalog number:  12347750

 Related applications: Antibody & Cell Culture Production | Protein Expression | Transfection | Viral Vaccine Production

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

293fectin™ Transfection Reagent is a proprietary, cationic lipid-based formulation for transfecting DNA into eukaryotic cells. This reagent is optimized for transfecting suspension 293 human embryonic kidney cells (e.g., FreeStyle™ 293-F cells, Cat. No. R790-07) in defined, serum-free FreeStyle™ 293 Expression Medium (Cat. No. 12338-018), and is intended for use with the FreeStyle™ 293 Expression System (Cat. No. K9000-01). 293fectin™ Transfection Reagent provides the following advantages:

• Demonstrates high transfection efficiency in suspension 293 cells and is also suitable for transfecting adherent 293 cells
• Suspension FreeStyle™ 293-F cells may be transfected in FreeStyle™ 293 Expression Medium; no medium change is required
• Add 293fectin™ reagent complexes directly to cells in culture medium
• No need to remove complexes or change or add medium following transfection
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Cell Type: Established Cell Lines
Sample Type (Specific): Plasmid DNA
Transfection Technique: Lipid-Based Transfection
Serum Compatible: Yes
Format: Tube(s)
High Throughput Compatibility: Not High Throughput-Compatible (Manual)
Product Line: 293fectin™
Product Size: 10 x 15 mL

Contents & storage

Each kit contains 10 units. Each unit contains one vial of reagent. Store at +4°C. 293fectin™ is guaranteed stable for 6 months when stored properly.

Documents

Manuals & protocols

Recommended products